Tisk…

(±)-Ethyl-2-methylbutyrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (±)-2-Methylbutanoic acid ethyl ester, (±)-2-Methylbutyric acid ethyl ester, Ethyl 2-methylbutanoate

W244309-1KG-K W244309-20KG-K W244309-4KG-K
SIALW244309-1KG-KEA 2680 CZK
SIALW244309-1KG-K SIALW244309-20KG-K SIALW244309-4KG-K
(±)-Ethyl-2-methylbutyrate, Sigma-Aldrich®
(±)-Ethyl-2-methylbutyrate
Vzorec: C₇H₁₄O₂
MW: 130.19
MDL Number: MFCD00012217
CAS číslo: 7452-79-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR