Tisk…

DL-Serin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid

M-Clarity™ quality level = MQ200

S5386-100G S5386-25G
SIALS5386-100GEA 2810 CZK
SIALS5386-100G SIALS5386-25G
DL-Serin, Sigma-Aldrich®
DL-Serin
Vzorec: C₃H₇NO₃
MW: 105.09
Bod varu: 395
Bod tání: 243…244 °C
Hustota: 1.543 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064223
CAS číslo: 302-84-1
EINECS: 206-130-6
Merck Index: 13,08534

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR