DL-Serin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid
M-Clarity™ quality level = MQ200
S4375-100G
S4375-1KG
SIALS4375-100GEA
4740
CZK
SIALS4375-100G
SIALS4375-1KG
DL-Serin, Sigma-Aldrich®
DL-Serin
Vzorec:
C₃H₇NO₃ MW: 105.09 Bod varu: 395 Bod tání: 243…244 °C Hustota: 1.543 Storage Temperature: Ambient |
MDL Number:
MFCD00064223 CAS číslo: 302-84-1 EINECS: 206-130-6 Merck Index: 13,08534 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...