Methyl (±)-lactate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Methyl Lactate, Methyl DL-lactate, Lactic acid methyll ester, Methyl (±)-2-hydroxypropionate
69822-25ML
69822-5ML
SIAL69822-25MLEA
6530
CZK
SIAL69822-25ML
SIAL69822-5ML
Methyl (±)-lactate, Sigma-Aldrich®
Methyl (±)-lactate
Vzorec:
C₄H₈O₃ Bod varu: 35 °C (1013 hPa) Hustota: 1,118 g/cm³ (0 °C) |
MDL Number:
MFCD00066367 CAS číslo: 547-64-8 EINECS: 208-930-0 UN: 3272 ADR: 3,III Merck Index: 12,06167 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...