Tisk…

Kyselina jodovodíková 55 - 58% ACS

Dodavatel: Thermo Fisher Scientific
Nebezpečí

36484.AD 36484.AP 36484.AY
36484.ADEA 2140 CZK
36484.AD 36484.AP 36484.AY
Kyselina jodovodíková 55 - 58% ACS
Kyselina jodovodíková

Suitable for use in methoxyl determinations.


Used as catalyst for the conversion of primary alcohol to alkyl halide.


Upozornení: Light sensitive liquid.

Vzorec: HI
Bod varu: 127 °C (1013 hPa)
Hustota: 1,701
Storage Temperature: Ambient
MDL Number: MFCD00011347
CAS číslo: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8,II
Merck Index: 14,4776

Objednat

Výsledky specifikačního testu

Assay (Unspecified) 55.0 - 58.0%
Residue on Ignition Max. 0.01%
Impurity Content, Chloride and Bromide (as Cl) Max. 0.05%
Impurity Content, Fe Max. 0.001%
Impurity Content, I₂ Max. 0.75%
Impurity Content, PO₄ Max. 0.001%
Impurity Content, SO₄ Max. 0.005%
Trace Metals (as Pb) Max. 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR