Tisk…

Orcein (synthetic), certified by the Biological Stain Commission, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Orcein (from synthetic), Natural Red 28

M-Clarity™ quality level = MQ200

O7505-25G O7505-5G
SIALO7505-25GEA 0 CZK
SIALO7505-25G SIALO7505-5G
Orcein (synthetic), certified by the Biological Stain Commission, Sigma-Aldrich®
Orcein

Certified for use in Darrow′s elastic fiber stain; Orcein method for hepatitis B surface antigen.

Storage Temperature: Room Temperature
MDL Number: MFCD00062310
CAS číslo: 1400-62-0
EINECS: 215-750-6

Objednat

Výsledky specifikačního testu

Appearance (Form) Powder
E1%
574 to 582 nm, 0.01 M NaOH
≥250
Wavelength 574 - 582 nm
Solubility (Colour) Purple to Dark Purple
Solubility (Turbidity)
1 mg/ml of 1 M NaOH
Clear
Biological Stain Commission
Dye Code Number
Pass
Recommended Retest Period 5 Years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR