Tisk…

Ethanol-[D₁], Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Hydroxy ethane-D1, Ethyl alcohol-D1, Ethanol-[²H₁], Methyl carbinol-D1

M-Clarity™ quality level = MQ100

452556-100ML 452556-25ML
SIAL452556-100MLEA 4990 CZK
SIAL452556-100ML SIAL452556-25ML
Ethanol-[D₁], Sigma-Aldrich®
Ethanol-[D₁]
Vzorec: C₂H₅DO
MW: 47.06
Bod varu: 78
Bod tání: –114.5 °C
Hustota: 0.826 g/cm³ (20 °C)
CAS číslo: 925-93-9
EINECS: 213-128-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR