měď ≥99.9% (metals basis), nanopowder APS 20-50 nm
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
45504.06
45504.14
45504.06EA
7310
CZK
45504.06
45504.14
měď ≥99.9% (metals basis), nanopowder APS 20-50 nm
měď
Purity: 99.9%
CAS No.: 7440-50-8
Molecular Formula: Cu(core)/CuO(shell)
Physical Form: CuO layer approx. 2nm
Appearance: Black
Odour: Odourless
UN No.: 3089
MDL No.: MFCD00010965
Vzorec:
Cu MW: 63.55 Bod varu: 2595 Bod tání: 1083 Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS číslo: 7440-50-8 EINECS: 231-159-6 UN: 3089 ADR: 4.1 II Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...