Selen, powder, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
229865-100G
229865-20G
229865-5G
SIAL229865-100GEA
24550
CZK
SIAL229865-100G
SIAL229865-20G
SIAL229865-5G
Selen, powder, Sigma-Aldrich®
Selenium
Vzorec:
Se Bod varu: 684,9 °C (1013 hPa) Bod tání: 217 °C Hustota: 4,81 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134090 CAS číslo: 7782-49-2 EINECS: 231-957-4 UN: 3283 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...