Tisk…

bromfenolová modř, AnalaR NORMAPUR® ACS

Dodavatel: VWR Chemicals

Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein

85721.050EA 1380 CZK
85721.050 85721.130
bromfenolová modř, AnalaR NORMAPUR® ACS
bromfenolová modř
Vzorec: C₁₉H₁₀Br₄O₅S
MDL Number: MFCD00005875
CAS číslo: 115-39-9

Objednat

Výsledky specifikačního testu

Clarity of solution Passes test ACS Characteristics Specifications
Identification (UV/VIS) Passes test
Sensitivity Passes test
Visual transition interval pH 3,0 - pH 4,6
Loss on drying (110 °C) Max. 2 %
Absorption max. λ 1 max (buffer pH 3,0) 434 - 439 nm
Absorption max. λ 2 max (buffer pH 4,6) 590 - 593 nm
Sp.absorptiv.(λ1max; 0,005g/l; pH3,0; dry) 350 - 400
Sp.absorptiv.(λ2max; 0.005g/l; pH4,6; dry) 940 - 1050


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR