Tisk…

(±)-Epichlorhydrin, GPR RECTAPUR®

Dodavatel: VWR Chemicals

Synonyms: (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane

87074.290EA 972 CZK
87074.290 87074.460
(±)-Epichlorhydrin, GPR RECTAPUR®
(±)-Epichlorhydrin
Vzorec: C₃H₅ClO
MW: 92,52 g/mol
MDL Number: MFCD00005132
CAS číslo: 106-89-8
Merck Index: 13,03642

Objednat

Výsledky specifikačního testu

Appearance Clear liquid
Assay (GC) ≥99.0 %
Density (d 20 °C/ 4 °C) 1.178 - 1.182
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR