Tisk…
Cut resistant gloves, Showa 542
Dodavatel: Showa
542-06 542-07 542-08 542-09 542-10
112-5866EA 6100 CZK
112-5866 112-5867 112-5868 112-5869 112-5870
Cut resistant gloves, Showa 542
Rukavice
These cut and abrasion resistant gloves made from HPPE protect hands during component assembly and hazardous applications
  • Polyurethane coating over a HPPE reinforced liner
  • Optimum flexibility and dexterity
  • Open back of hand design reduces perspiration
  • Seamless liner reduces irritation during long periods of wear
  • Elasticated wrist cuff
These gloves were made with high performance Poly Ethylene (HPPE), coated palm, elasticated cuff, standard finish and ideally suited for aerospace, automotive, construction, manufacturing, ceramics, steel and metals. Typical application includes electrical appliance assembly, handling metal parts, precision mechanics, product protection, white goods handling and wire assembly.
Certifikace: EN 388:2016 (4341B); Cat. II
Balení: 10 pairs per polybag.