Isopropyl myristate ≥98%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Synonyms:
Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester, Isopropyl Myristate, Isopropyl tetradecanoate, Tetradecylic acid isopropyl ester
85991.180EA
327
CZK
85991.180
85991.290
85991.320
Isopropyl myristate ≥98%, GPR RECTAPUR®
Isopropyl myristate
Vzorec:
C₁₇H₃₄O₂ MW: 270,46 g/mol |
MDL Number:
MFCD00008982 CAS číslo: 110-27-0 |
Výsledky specifikačního testu
Assay | Min. 98 % |
Density (20/4) | About 0.853 |
Acid value | Max. 0,5 |
Iodine value (g/100 g) | Max. 1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...