Tisk…
ESD safety shoes, lace-up, uvex 2 trend, 6947
Dodavatel: UVEX
6947.8/44 6947.8/45 6947.8/46 6947.8/47 6947.8/48 6947.8/49 6947.8/50 6947.8/51 6947.8/52 6947.9/38 6947.9/39 6947.9/40 6947.9/41 6947.9/42 6947.9/43 6947.9/44 6947.9/45 6947.9/46 6947.9/47 6947.9/48 6947.9/49 6947.9/50 6947.9/51 6947.9/52 6947.7/38 6947.7/39 6947.7/40 6947.7/41 6947.7/42 6947.7/43 6947.7/44 6947.7/45 6947.7/46 6947.7/47 6947.7/48 6947.7/49 6947.7/50 6947.7/51 6947.7/52 6947.8/38 6947.8/39 6947.8/40 6947.8/41 6947.8/42 6947.8/43
UVEX6947.8/44EA 2270 CZK
UVEX6947.8/44 UVEX6947.8/45 UVEX6947.8/46 UVEX6947.8/47 UVEX6947.8/48 UVEX6947.8/49 UVEX6947.8/50 UVEX6947.8/51 UVEX6947.8/52 UVEX6947.9/38 UVEX6947.9/39 UVEX6947.9/40 UVEX6947.9/41 UVEX6947.9/42 UVEX6947.9/43 UVEX6947.9/44 UVEX6947.9/45 UVEX6947.9/46 UVEX6947.9/47 UVEX6947.9/48 UVEX6947.9/49 UVEX6947.9/50 UVEX6947.9/51 UVEX6947.9/52 UVEX6947.7/38 UVEX6947.7/39 UVEX6947.7/40 UVEX6947.7/41 UVEX6947.7/42 UVEX6947.7/43 UVEX6947.7/44 UVEX6947.7/45 UVEX6947.7/46 UVEX6947.7/47 UVEX6947.7/48 UVEX6947.7/49 UVEX6947.7/50 UVEX6947.7/51 UVEX6947.7/52 UVEX6947.8/38 UVEX6947.8/39 UVEX6947.8/40 UVEX6947.8/41 UVEX6947.8/42 UVEX6947.8/43
ESD safety shoes, lace-up, uvex 2 trend, 6947
Obuv
Sporty S1 safety shoel with wide fit.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Breathable microvelour upper
  • Penetration resistant steel midsole and steel toe cap
  • Reflective details for enhanced visibility
Safety Shoes feature an ergonomically designed outsole that offers excellent slip resistance, as well as a stabilising uvex anti-twist heel cap designed to prevent ankle twisting. Suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
Colour: Black/grey/orange
Certifikace: EN 61340 (ESD); EN ISO 20345:2011, S1, SRC