Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ≥95.0% (ICP, K basis), Selectophore™ ionophore, Supelco®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
60588-500MG
60588-50MG
60588-500MGEA
63570
CZK
60588-500MG
60588-50MG
Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ≥95.0% (ICP, K basis), Selectophore™ ionophore, Supelco®
Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate
Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate was used as one of the membrane component during preparing ion-selective electrodes (ISEs), in an experimental study conducted to classify beverages using a reduced sensor array.
Balení: Bottomless glass bottle. Contents are inside inserted fused cone.
Vzorec:
C₃₂H₁₂BF₂₄K |
MDL Number:
MFCD00077934 CAS číslo: 105560-52-9 |
|
Výsledky specifikačního testu
Appearance (Color) | White to Faint Beige |
Thin Layer Chromatography | Corresponds to Standard |
Proton NMR Spectrum | Conforms to Structure |
Assay (ICP) | ≥ 95.0 % Determined by ICP (Based on K) |
Recommended Retest Period | 54 Months |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...