Tisk…
Nitrate Ionophore VI, Selectophore™, function tested ionophore, Supelco®
Dodavatel: Merck
Synonyms: 9-Hexadecyl-1,7,11,17-tetraoxa-2,6,12,16-tetraazacycloeicosane
07295-100MG 07295-10MG 07295-500MG
07295-100MGEA 33260 CZK
07295-100MG 07295-10MG 07295-500MG
Nitrate Ionophore VI, Selectophore™, function tested ionophore, Supelco®
Nitrate Ionophore VI
Ionophore for online monitoring of nitrate obtains superior characteristics over all existing nitrate ionophores.
Nitrate ionophore VI may be used in polyvinyl chloride/dioctylsebacate (PVC/DOS) matrix using a highly sensitive piezoresitive silicon oxide cantilevers for determination of nitrates in soil.
Balení: Bottomless glass bottle. Contents are inside inserted fused cone
Vzorec: C₂₈H₆₀N₄O₄
MW: 516.80 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16652520
CAS číslo: 1196157-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52148a998ee9948ae7d4c569ea8ecfd15b38067e&vid=1953d8c4be3bff497aab3e6e4464279b68e8246e&prid=web&clid=SAPDAM