Bis(1-butylpentyl) adipate ≥98.0%, Selectophore™ ionophore, Supelco®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
02150-25ML
02150-5ML
02150-25MLEA
16570
CZK
02150-25ML
02150-5ML
Bis(1-butylpentyl) adipate ≥98.0%, Selectophore™ ionophore, Supelco®
Bis(1-butylpentyl) adipate
Plasticizer employed in polymeric membrane electrodes.
Vzorec:
C₂₄H₄₆O₄ Bod vzplanutí: 110 °C |
MDL Number:
MFCD00042947 CAS číslo: 77916-77-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...