Zincon sodium salt for spectrophotometric determination of Cu, for the determination of zinc, Supelco®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
96440-1G
96440-5G
96440-1GEA
2770
CZK
96440-1G
96440-5G
Zincon sodium salt for spectrophotometric determination of Cu, for the determination of zinc, Supelco®
Zincon sodium salt
A colorimetric indicator for Zn, Cu and Hg
Vzorec:
C₂₀H₁₅N₄NaO₆S |
MDL Number:
MFCD00064385 CAS číslo: 62625-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...