Amberlite® IRA743 free base ion-exchange resin, Supelco®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
216445-1KG
216445-250G
216445-1KGEA
10020
CZK
216445-1KG
216445-250G
Amberlite® IRA743 free base ion-exchange resin, Supelco®
Amberlite® IRA743
Amberlite® IRA743 free base is of styrenic, macroreticular form. Weakly basic resin with N-methylglucaomine functionality which is useful in boron-specific applications.
Borate-specific chelating resin for the removal of borate, boric acid, other boron species from water; highly selective so that salts, including bases, do not interfere significantly.
MDL Number:
MFCD00132701 CAS číslo: 63181-98-6 |