Tisk…
Nucleosil 100 diol, HPLC Columns
Dodavatel: Avantor
NC100-7DIOL-50AEA 12740 CZK
NC100-7DIOL-50A
Nucleosil 100 diol, HPLC Columns
Kolony chromatografické
Packed with classical spherical porous silica available in a wide range of pore sizes. The 100 and 120 Å have similar relative retention characteristics, however the 100 Å has the greater capacity.
  • Good mechanical stability
  • Good batch to batch reproducibility