Tisk…

Hematoxylin (Mayer) for histology, J.T.Baker®

Dodavatel: Avantor

3870.2500EA 2970 CZK
3870.2500 3870.1000
Hematoxylin (Mayer) for histology, J.T.Baker®
Hematoxylin

Hematoxyline solution according to mayer for use in HE staining.

Vzorec: C₁₆H₁₄O₆
MW: 302,28 g/mol
MDL Number: MFCD00078111
CAS číslo: 517-28-2
IVDLogo

Objednat

Výsledky specifikačního testu

Performance Test Passes Test
For Laboratory, Research or Manufacturing Use

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR