Eosin 225 for histology
Dodavatel: RAL (REACTIFS RAL SA)
|
Varování
|
720-0178EA
2110
CZK
720-0178
Eosin 225 for histology
Eozin Y
Vzorec:
C₂₀H₆Br₄Na₂O₅ MW: 691,86 g/mol |
MDL Number:
MFCD00005040 CAS číslo: 17372-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...