1-Octanesulphonic acid sodium salt monohydrate ≥99.0% (by titrimetric analysis), Supelco®
Dodavatel: Merck
Synonyms:
Sodium 1-octansulphonate monohydrate, Sodium 1-octanesulphonate monohydrate
M-Clarity™ quality level = MQ100
74884-10G
74884-5G
74884-25G
74884-10GEA
6620
CZK
74884-10G
74884-5G
74884-25G
1-Octanesulphonic acid sodium salt monohydrate ≥99.0% (by titrimetric analysis), Supelco®
1-Octanesulphonic acid sodium salt monohydrate
Vzorec:
C₈H₁₇NaO₃S·1H₂O |
MDL Number:
MFCD00007544 CAS číslo: 207596-29-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...