N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide with 1% tert-butylchlorodimethylsilane ≥95.0% (by GC), LiChropur™ for GC derivatisation, Supelco®
Dodavatel: Merck
|
Varování
|
Synonyms:
N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide, Silylating mixture VI, MTBSTFA + 1% TBDMSCl
00942-10X1ML
00942-10ML
00942-10X1MLEA
13630
CZK
00942-10X1ML
00942-10ML
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide with 1% tert-butylchlorodimethylsilane ≥95.0% (by GC), LiChropur™ for GC derivatisation, Supelco®
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide with 1% tert-butylchlorodimethylsilane
Vzorec:
C₉H₁₈F₃NOSi MW: 241,33 g/mol |
MDL Number:
MFCD00009671 CAS číslo: 77377-52-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...