Tisk…
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane 99% (excluding TMCS) (by GC), LiChropur™ for GC derivatisation, Supelco®
Dodavatel: Merck
Nebezpečí
Synonyms: BSTFA+TMCS (99:1)
15238-100ML 15238-25ML 15238-5ML 15238-10X1ML 15238-10X0.1ML 15238-1ML
15238-100MLEA 19390 CZK
15238-100ML 15238-25ML 15238-5ML 15238-10X1ML 15238-10X0.1ML 15238-1ML
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane 99% (excluding TMCS) (by GC), LiChropur™ for GC derivatisation, Supelco®
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Vzorec: C₈H₁₈F₃NOSi₂
MW: 257.4
Storage Temperature: Freezer
MDL Number: MFCD00008269
CAS číslo: 25561-30-2
UN: 2924
ADR: 8(3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a306c5840c61512f9d0f83bc8d9bda47ac488b9f&vid=03b1f13daeb83c089b7ffd977bd00bcc5f159fd9&prid=web&clid=SAPDAM