Tisk…
N,N-Dietyl-p-fenylendiamoniumsulfát ∼98% Min.
Dodavatel: Spectrum Chemical
Varování
Synonyms: N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, 4-Amino-N,N-diethylaniline sulphate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate
D3427-25KGBL D3427-5KGBL
SPCMD3427-25KGBLEA 0 CZK
SPCMD3427-25KGBL SPCMD3427-5KGBL
N,N-Dietyl-p-fenylendiamoniumsulfát ∼98% Min.
N,N-Dietyl-p-fenylendiamoniumsulfát
N,N-Diethyl-p-phenylenediamine sulphate has applications in the laboratory including for the spectrophotometric determination of phenolic and aromatic amino groups. It has also been used as a reference probe to study rapid chlorination rate constants by a stopped-flow spectrophotometric competition kinetics method
Vzorec: C₁₀H₁₈N₂O₄S
MW: 262,33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012993
CAS číslo: 6283-63-2
Objednat
Výsledky specifikačního testu
Assay ∼98% Min.