Tisk…
Peaked caps
Dodavatel: VWR Collection
113-9236EA 0 CZK
113-9236 113-9237 113-9238 113-9239
Peaked caps
Jednorázové oděvy
These disposable caps, with peak, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in white and blue
Weight: 40 g/m² (top), 100 g/m² (peak)
Balení: 100 pieces per polybag and 1000 pieces per carton.