Tisk…

Kyanokobalamin ≥95%, crystalline solid

Dodavatel: Cayman Chemical
Varování

18425-1 18425-10 18425-5 18425-500
CAYM18425-1EA 2330 CZK
CAYM18425-1 CAYM18425-10 CAYM18425-5 CAYM18425-500
Kyanokobalamin ≥95%, crystalline solid
Kyanokobalamin

Vitamin B12 is a water-soluble vitamin with roles in brain and nervous system functioning and blood formation through regulation of DNA synthesis, cellular metabolism, fatty acid metabolism, and amino acid metabolism.


Vitamin B12 belongs to the corrinoid group of compounds, which contain a corrin macrocycle, and is produced only by certain bacteria and archaea. Reference to vitamin B12 usually encompasses a group of cobalt-containing compounds (or cobalamins), which include cyanocobalamin, hydroxocobalamin, 5'-deoxyadenosylcobalamin (AdoB12), and methylcobalamin (MeB12). These forms serve as cofactors in isomerase-, methyltransferase-, andehalogenase-catalysed reactions.

Vzorec: C₆₃H₈₈CoN₁₄O₁₄P
Storage Temperature: Freezer
MDL Number: MFCD00151092
CAS číslo: 68-19-9

Objednat

Výsledky specifikačního testu

Purity ≥95%
Formulation A Crystalline Solid
λmax 208, 278, 360, 548 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR