Tisk…

Tergitol® TMN-10 surfactant

Dodavatel: Spectrum Chemical

Synonyms: Isolaureth-10, Polyethylene glycol trimethylnonyl ether

T1288-500ML T1288-20LT T1288-2.5LTPL
SPCMT1288-500MLEA 8400 CZK
SPCMT1288-500ML SPCMT1288-20LT SPCMT1288-2.5LTPL
Tergitol® TMN-10 surfactant
Tergitol® TMN-10

Tergitol(R) TMN-10 (90% Aqueous), surfactant is a nonylphenol ethoxylate. It is non-ionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odour.


Used in cleaners & degreasers, prewash spotters and oil field chemicals.

Vzorec: C₁₂H₂₅O(CH₂CH₂O)nH
MW: ∼ 540 g/mol
MDL Number: MFCD00132411
CAS číslo: 60828-78-6

Objednat

Výsledky specifikačního testu

Cloud Point of a 1% Aqueous Solution 72 - 80 °C
pH of a 10% Solution at 25 °C 5.0 - 8.0
Water 8 - 10%
Suspended Matter Substantially Free
Color (APHA) Max. 200


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR