Tisk…

Chloramfenikol USP

Dodavatel: G Bioscienes
Nebezpečí

Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol

RC-168 RC-167
GENORC-168EA 3680 CZK
GENORC-168 GENORC-167
Chloramfenikol USP
Chloramfenikol

Tested against both sensitive and resistant cells.

Vzorec: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MDL Number: MFCD00078159
CAS číslo: 56-75-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR