Pure water density standard, Supelco®
Dodavatel: Merck
DENWAT3-3X60ML
DENWAT-60ML
SIALDENWAT3-3X60MLEA
15530
CZK
SIALDENWAT3-3X60ML
SIALDENWAT-60ML
Pure water density standard, Supelco®
Standardy
Standardy hustotní
Pure water density standard is basically designed for the verification or calibration of instruments used to measure density.
- n20/D = 1.34 (lit.)
- 0,9982 g/ml at 20 °C
- 0,9970 g/ml at 25 °C
- 1,000 g/ml at 3,98 °C (lit.)