Tisk…

Petunidin chloride ≥98%

Dodavatel: Cayman Chemical

Synonyms: 3,5,73',3'-Pentahydroxy-5'-methoxyflavylium chloride, 2-(3,4-Dihydroxy-5-methoxy-phenyl)-3,5,7-trihydroxy-chromenylium chloride, Myrtillidin chloride, flavyliuchloride

19755-1 19755-10 19755-25 19755-5
CAYM19755-1EA 2240 CZK
CAYM19755-1 CAYM19755-10 CAYM19755-25 CAYM19755-5
Petunidin chloride ≥98%
Petunidin chloride

Petunidin is an O-methylated anthocyanidin derived from delphinidin that imparts blue-red pigments to flowers, fruits, and red wine. Used to bind with and suppress the activity of focal adhesion kinase and to inhibit platelet-derived growth factor-induced aortic smooth muscle cell migration, which may confer a protective effect against atherosclerosis. Extracts from E. jambolana fruit, which is rich in anthocyanins including petunidin, have been used to suppress the proliferation of an HCT116 colon cancer cell line, as well as colon cancer stem cells.

Vzorec: C₁₆H₁₃O₇·Cl
Storage Temperature: Freezer
MDL Number: MFCD00017580
CAS číslo: 1429-30-7

Objednat

Výsledky specifikačního testu

Purity ≥98%
Formulation A Crystalline Solid
λmax 573 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR