Biotin hydrazide ≥95%
Dodavatel: Cayman Chemical
Synonyms:
5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide
Biotin hydrazide is a biotinyl derivative used to label surface functional groups, antibodies, lectins, sugars, nucleic acids, or molecules with free carboxylic or keto groups.
Used as a probe for determining protein carbonylation, an irreversible posttranslational modification resulting from the actions of reactive oxygen species or lipid oxidation products. The reaction of this probe is direct and does not require catalysts or reducing agents.
Vzorec:
C₁₀H₁₈N₄O₂S MW: 258.34 g/mol |
MDL Number:
MFCD00078532 CAS číslo: 66640-86-6 |
Výsledky specifikačního testu
Purity | ≥95% |
Formulation | A Crystalline Solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...