Tisk…

Coenzyme A sodium salt hydrate ≥95%

Dodavatel: Cayman Chemical

21722-10 21722-25 21722-5 21722-50
CAYM21722-10EA 2210 CZK
CAYM21722-10 CAYM21722-25 CAYM21722-5 CAYM21722-50
Coenzyme A sodium salt hydrate ≥95%
Coenzyme A sodium salt hydrate

Coenzyme A (CoA) is an essential cofactor functioning as an acyl group carrier and carbonyl-activating group for the citric acid cycle and fatty acid metabolism. About 4% of cellular enzymes utilize CoA as a substrate. It is synthesized from pantothenic acid in a five-step process that requires ATP. The pantothenate kinase step of the CoA biosynthetic pathway is identified as a target for the development of antibacterial compounds.

Vzorec: C₂₁H₃₆N₇O₁₆P₃S·xNa⁺·yH₂O
Storage Temperature: Freezer
MDL Number: MFCD06795857
CAS číslo: 55672-92-9

Objednat

Výsledky specifikačního testu

Purity ≥95%
Formulation A Crystalline Solid
λmax 258 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR