EPZ-5676 (Pinometostat) ≥98%
Dodavatel: Cayman Chemical
Synonyms:
(2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(((cis-3-(2-(5-(tert-butyl)-1H-benzo[d]imidazol-2-yl)ethyl)cyclobutyl)(isopropyl)amino)methyl)tetrahydrofuran-3,4-diol, Pinometostat
16175-10
16175-1
16175-25
16175-5
CAYM16175-10EA
11310
CZK
CAYM16175-10
CAYM16175-1
CAYM16175-25
CAYM16175-5
EPZ-5676 (Pinometostat) ≥98%
Pinometostat (EPZ-5676)
EPZ5676 is a highly potent aminonucleoside inhibitor of DOT1 l histone methyltransferase activity (Ki = 80 pM) that demonstrates 37000-fold selectivity over other methyltransferases.
It can selectively inhibit the proliferation of acute leukemia cell lines bearing MLL translocations (IC50s range from 3.5 nM - 1.3 µM).
Vzorec:
C₃₀H₄₂N₈O₃ MW: 562,71 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD24849415 CAS číslo: 1380288-87-8 |
Výsledky specifikačního testu
Purity | ≥98% |
Formulation | A Crystalline Solid |
λmax | 255 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...