Tisk…
ESD safety shoes, lace-up, uvex 2, Xenova®, 9555
Dodavatel: UVEX
9555.7/38 9555.7/39 9555.7/40 9555.7/41 9555.7/42 9555.7/43 9555.7/44 9555.7/45 9555.7/46 9555.7/47 9555.7/48 9555.7/49 9555.7/50 9555.7/51 9555.7/52 9555.8/38 9555.8/39 9555.9/52 9555.8/40 9555.8/41 9555.8/42 9555.8/43 9555.8/44 9555.8/45 9555.8/46 9555.8/47 9555.8/48 9555.8/49 9555.8/50 9555.8/51 9555.8/52 9555.9/38 9555.9/39 9555.9/40 9555.9/41 9555.9/42 9555.9/43 9555.9/44 9555.9/45 9555.9/46 9555.9/47 9555.9/48 9555.9/49 9555.9/50 9555.9/51
UVEX9555.7/38EA 3410 CZK
UVEX9555.7/38 UVEX9555.7/39 UVEX9555.7/40 UVEX9555.7/41 UVEX9555.7/42 UVEX9555.7/43 UVEX9555.7/44 UVEX9555.7/45 UVEX9555.7/46 UVEX9555.7/47 UVEX9555.7/48 UVEX9555.7/49 UVEX9555.7/50 UVEX9555.7/51 UVEX9555.7/52 UVEX9555.8/38 UVEX9555.8/39 UVEX9555.9/52 UVEX9555.8/40 UVEX9555.8/41 UVEX9555.8/42 UVEX9555.8/43 UVEX9555.8/44 UVEX9555.8/45 UVEX9555.8/46 UVEX9555.8/47 UVEX9555.8/48 UVEX9555.8/49 UVEX9555.8/50 UVEX9555.8/51 UVEX9555.8/52 UVEX9555.9/38 UVEX9555.9/39 UVEX9555.9/40 UVEX9555.9/41 UVEX9555.9/42 UVEX9555.9/43 UVEX9555.9/44 UVEX9555.9/45 UVEX9555.9/46 UVEX9555.9/47 UVEX9555.9/48 UVEX9555.9/49 UVEX9555.9/50 UVEX9555.9/51
ESD safety shoes, lace-up, uvex 2, Xenova®, 9555
Obuv
Very lightweight water repellent uvex 2 xenova® shoes are made from full-grain smooth leather upper, soft padded collar and a slip-resistant Xenova® dual-density PU sole resistant to heat, chemicals, oils and gasoline. All sole materials are free of silicones, plasticisers and other paint-wetting impairment substances and suitable for people allergic to chrome.
  • Compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
  • Anti-twist rear cap provides more stability and kink protection
  • Climazone technology for improved comfort
  • Anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Heat-resistant loop lacing system
  • DUO-PUR sole, non-marking, with excellent cushioning and abrasion-resistant outsole
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Shock absorbing heel and forefoot
  • uvex xenova® composite toe cap (200 J)
  • Fulfils the ESD requirement: Discharge resistance <35 megohm
Used for medium-duty applications.
Colour: Blue/black
Certifikace: EN 61340 (ESD); EN ISO 20345:2011, S2, SRC