Tisk…
ESD safety ankle boots, lace-up, Mason Pro Rubber Mid
Dodavatel: ELTEN
7682101/40 7682101/41 7682101/42 7682101/43 7682101/44 7682101/45 7682101/46 7682101/47 7682101/48 7682102/40 7682102/41 7682102/42 7682102/43 7682102/44 7682102/45 7682102/46 7682102/47 7682102/48 7682103/40 7682103/41 7682103/42 7682103/43 7682103/44 7682103/45 7682103/46 7682103/47 7682103/48
ELTN7682101/40EA 2700 CZK
ELTN7682101/40 ELTN7682101/41 ELTN7682101/42 ELTN7682101/43 ELTN7682101/44 ELTN7682101/45 ELTN7682101/46 ELTN7682101/47 ELTN7682101/48 ELTN7682102/40 ELTN7682102/41 ELTN7682102/42 ELTN7682102/43 ELTN7682102/44 ELTN7682102/45 ELTN7682102/46 ELTN7682102/47 ELTN7682102/48 ELTN7682103/40 ELTN7682103/41 ELTN7682103/42 ELTN7682103/43 ELTN7682103/44 ELTN7682103/45 ELTN7682103/46 ELTN7682103/47 ELTN7682103/48
ESD safety ankle boots, lace-up, Mason Pro Rubber Mid
Obuv
ESD safety ankle boots made from wear-resistant breathable cowhide leather with fabric lining, reflective materials, a full padded bellows tongue and a dual density rubber/PU sole ERGO-ACTIVE 2.
  • Fabric lining for natural ventilation
  • Replaceable insole; orthopaedic insoles possible
  • Semi-orthopaedic inlay sole ESD black
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • TPU scuff cap
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Type 1: Suitable for larger and strong feet, short toes with wide ball, heel area and steep ball angle.
Type 2: Suitable for normal feet, long toes with medium-wide ball, heel area and flat ball angle.
Type 3: Suitable for slim feet, medium-sized toes with narrow ball, heel area and medium ball angle.
Certifikace: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, form B