Phorbol-12-myristate-13-acetate 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
TPA, Phorbol 12-myristate 13-acetate, 12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, (1aR,1bS,4aR,7aS,7bS,8R,9R,9aS)-9a-Acetoxy-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1a,1b,4,4a,5,7a,7b,8,9,9a-decahydro-1H-cyclopropa[3,4]benzo[1,2-e]azulen-9-yl tetradecanoate
PMA activates Ca2+ - ATPase and potentiates forskolin-induced cAMP formation. Used to inhibit apoptosis induced by the Fas antigen, but PMA induces apoptosis in HL-60 promyelocytic leukemia cells.
Very soluble in methanol, soluble in DMSO, chloroform, ethanol or acetone, ether, ethyl acetate, DMF or acetonitrile. Insoluble in water.
Upozornení: Light sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Vzorec:
C₃₆H₅₆O₈ MW: 616,836 g/mol |
MDL Number:
MFCD00036736 CAS číslo: 16561-29-8 |
Výsledky specifikačního testu
Appearance (Color) | White to pale cream |
Form | Powder, film or foam, solid |
Assay from Suppliers CofA | ≥94.0% |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...