Tisk…
Aluminium oxide activated basic Brockmann Grade I ~150 mesh, Fluka™
Dodavatel: Honeywell Chemicals
199443-1KG 199443-5KG 199443-20KG
199443-1KGEA 3640 CZK
199443-1KG 199443-5KG 199443-20KG
Aluminium oxide activated basic Brockmann Grade I ~150 mesh, Fluka™
Aluminium oxide activated basic
Vzorec: Al₂O₃
MW: 101.96
Bod varu: 2.980 °C (1.013 hPa)
Bod tání: 2.072 °C
Hustota: 3.960 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS číslo: 1344-28-1
EINECS: 215-691-6
Objednat
Výsledky specifikačního testu
Brockmann Number (Activity) I (val.ref) complying
pH (10 % susp., 20°C) 8.5 - 10.5
Spez. surface (BET) 184.5 - 225.5 m²/g
Fe (as Fe₂O₃) Max. 0.03 %
Na (as Na₂O) Max. 0.4 %
Silicon (as SiO₂) Max. 0.03 %
Chloride (Cl) complying
Grain size 0.04-0.30 mm Min. 95 %
Form powder