Tisk…
Cell culture media, Dulbecco’s modified eagle’s medium (DMEM), high glucose
Dodavatel: Merck
D6429-1L D6429-24X500ML D6429-500ML D6429-6X1L D6429-6X500ML
SIALD6429-1LEA 1990 CZK
SIALD6429-1L SIALD6429-24X500ML SIALD6429-500ML SIALD6429-6X1L SIALD6429-6X500ML
Cell culture media, Dulbecco’s modified eagle’s medium (DMEM), high glucose
Média kultivační
Dulbecco′s modified eagle′s medium (DMEM) is a modification of basal medium eagle (BME) that contains four-fold concentrations of the amino acids and vitamins.
This DMEM-High glucose medium is a 1X complete medium with sodium pyruvate added. It also differs from the original DMEM-High formulation wherein pyridoxine is substituted for pyridoxal. Pyridoxal is an unstable component of media.