Tisk…
FLUO 3/AM ≥95% (by HPLC) fluorescent indicator, for calcium determination
Dodavatel: Biotium
Synonyms: 4-(6-Acetoxymethoxy-2,7-dichloro-3-oxo-9-xanthenyl)-4′-methyl-2,2′(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, 1-[Amino-5-(2,7-dichloro-6-acetomethoxy-3-oxo-3H-xanthen-9-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid pentaacetoxy-methyl ester
50013 50014 50016
50013.EA 12210 CZK
50013. 50014. 50016.
FLUO 3/AM ≥95% (by HPLC) fluorescent indicator, for calcium determination
FLUO 3/AM
Membrane-permeant AM ester form of Fluo-3 that can be loaded into cells via incubation. Fluo-3 AM ester itself does not bind Ca2 , but it is readily hydrolyzed to Fluo-3 by endogenous esterases once the dye is inside the cells. An important application of Fluo-3 AM ester is its use in high throughput drug screening.
  • Abs/Em (after hydrolysis): 506/526 nm (low or high [Ca2 ] )
  • Extinction Coefficient (after hydrolysis): 86,000 M-1 cm-1 (506 nm)
  • Kd (Ca2 ) (after hydrolysis): 450 nM
  • Cell permeability: Membrane permeant
  • Orange red solid soluble in DMSO
  • Store -20°C and protect from light, especially when in solution
Because of the relatively low water solubility of the AM ester, Pluronic F-127, a mild detergent, is often used as a dispersing agent to facilitate dye loading. See the product information sheet for more information.
Vzorec: C₅₁H₅₀Cl₂N₂O₂₃
MW: 1129,85 g/mol
Storage Temperature: Freezer
CAS číslo: 121714-22-5