Tisk…
Heating mantle, KM-MPE series
Dodavatel: MOHR
83001000 93003000
MOHR83001000EA 24820 CZK
MOHR83001000 MOHR93003000
Heating mantle, KM-MPE series
Ohřívače Hnízda topná
Metal-cased heating mantle with built-in controller for operating temperatures up to 450 °C.
  • Flexible glass yarn heating element provides even heat transfer and gentle treatment of the glassware
  • Chemical resistant metal housing
  • Thermally insulated and earthed
  • Predrilled threaded hole for clamp KM-SK
  • Built-in heating zone switch
System of protection: Casing IP 43, heating surface IP 00.