Tisk…
Coats, paint-tex® plus
Dodavatel: Zellstoff-Vertriebs-GmbH & Co. KG
30773-00 30775-00 30774-00
113-9160EA 20100 CZK
113-9160 ZVGA30775-00 ZVGA30774-00
Coats, paint-tex® plus
Jednorázové oděvy
Suitable for longer use due to the breathable SMS material.
  • Features snap buttons
  • Collar and cuffs
  • Three pockets
Ideal for renovation work, in the remediation of contaminated sites and asbestos.
Certifikace: Cat. III/CE 1023/Type 5 + 6