Tisk…
Heavy duty hoses for airhoods
Dodavatel: Honeywell Safety Products
NORHA161253EA 19930 CZK
NORHA161253
Heavy duty hoses for airhoods
Hadičky Hadičky vakuové/tlakové
Black heavy duty breathing hoses for use in connection with light and heavy duty airhoods.
  • Anti-static
  • With quick connection double action safety male and female couplings
  • Available in several lengths
Certifikace: EN 139