Tisk…
Racks for freezers
Dodavatel: EPPENDORF
6001050.111 6001000.310 6001000.311 6001000.410 6001000.411 6001000.510 6001000.910 6001000.911 6001011.110 6001011.210 6001011.310 6001011.410 6001011.510 6001011.910 6001012.210 6001012.310 6001012.410 6001012.510 6001012.910 6001021.110 6001021.210 6001021.310 6001021.410 6001021.910 6001022.210 6001022.310 6001022.410 6001022.910 6001040.010 6001040.011 6001040.110 6001040.111 6001040.210 6001040.211 6001050.010 6001050.011 6001050.110 6001050.210 6001050.211 6001061.210 6001061.310 6001061.410 6001061.510 6001061.910 6001062.210 6001062.310 6001062.410 6001062.510 6001062.910 6001072.210 6001082.210 6001071.110 6001071.210 6001071.310 6001071.410 6001071.510 6001071.910 6001072.310 6001072.410 6001072.510 6001072.910 6001081.110 6001081.210 6001081.310 6001081.410 6001081.510 6001081.910 6001082.310 6001082.410 6001082.510 6001082.910
EPPE6001050.111EA 0 CZK
EPPE6001050.111 EPPE6001000.310 EPPE6001000.311 EPPE6001000.410 EPPE6001000.411 EPPE6001000.510 EPPE6001000.910 EPPE6001000.911 EPPE6001011.110 EPPE6001011.210 EPPE6001011.310 EPPE6001011.410 EPPE6001011.510 EPPE6001011.910 EPPE6001012.210 EPPE6001012.310 EPPE6001012.410 EPPE6001012.510 EPPE6001012.910 EPPE6001021.110 EPPE6001021.210 EPPE6001021.310 EPPE6001021.410 EPPE6001021.910 471-1209 EPPE6001022.310 EPPE6001022.410 EPPE6001022.910 EPPE6001040.010 EPPE6001040.011 EPPE6001040.110 EPPE6001040.111 EPPE6001040.210 EPPE6001040.211 EPPE6001050.010 EPPE6001050.011 EPPE6001050.110 EPPE6001050.210 EPPE6001050.211 EPPE6001061.210 EPPE6001061.310 EPPE6001061.410 EPPE6001061.510 EPPE6001061.910 EPPE6001062.210 EPPE6001062.310 EPPE6001062.410 EPPE6001062.510 EPPE6001062.910 471-1224 471-1225 EPPE6001071.110 EPPE6001071.210 EPPE6001071.310 EPPE6001071.410 EPPE6001071.510 EPPE6001071.910 EPPE6001072.310 EPPE6001072.410 EPPE6001072.510 EPPE6001072.910 EPPE6001081.110 EPPE6001081.210 EPPE6001081.310 EPPE6001081.410 EPPE6001081.510 EPPE6001081.910 EPPE6001082.310 EPPE6001082.410 EPPE6001082.510 EPPE6001082.910
Racks for freezers
Stojany Poličky pro kryoboxy
These racks are heavy-duty and designed for higher capacity and durability. Specially designed for optimised cooling conditions and improved sample safety.
  • Fit freezer boxes up to a 136×136 mm for increased storage flexibility
  • Drawer-style freezer racks, each drawer can be individually removed, leaving the rest in the freezer
  • In side-access racks each box fits into a distinct, individual compartment for easier inventory tracking
There are two systems used in laboratories - the standard 4-boxes deep racks and 5-boxes deep racks. The easiest way is to take out all racks (with the samples/ boxes included) and store those racks in some backup freezer. To be compatible with most common ULT freezer suppliers, these ULT freezers are designed for 4-boxes deep racks.
The ULT freezer with 3 compartments/shelves has higher compartments compared to the 5-compartment ULT freezer. Racks of a 5-compartment freezer do fit into a 3-compartment freezer but not vice versa. Most of the storage boxes have to be pulled out of the in the cold inner area of the ULT freezer to access the samples.
To get the maximum capacity out of the ULT freezer, the CryoCube F740 series (3-compartment) can be equipped with 3-compartment MAX racks. These racks only fit into the CryoCube F740 series 3-compartment instruments.
Informace o dodávce: Locking rods are now available for chest freezer racks to provide additional sample security.