Tisk…
Certified vial kits, Supelco®, Sigma-Aldrich
Dodavatel: Merck
29378-U 29379-U 29381-U 29384-U 29385-U 29386-U 29387-U 29403-U 29432-U
SIAL29378-UEA 1930 CZK
SIAL29378-U SIAL29379-U SIAL29381-U SIAL29384-U SIAL29385-U SIAL29386-U SIAL29387-U SIAL29403-U SIAL29432-U
Certified vial kits, Supelco®, Sigma-Aldrich
Vialky
These certified vial kits are designed, engineered, and manufactured for optimal performance in HPLC, GCMS, and LCMS. The vial, cap, and septa are batch tested to confirm that the quality of each component will be the same, time after time.
  • These vials work with all autosamplers and have the quality and reproducibility to ensure a successful analysis
  • Offered in a variety of styles such as 9 mm thread, crimp top and snap top vials
Vials made of type 1 borosilicate glass. Tight tolerances on all physical specifications are controlled to make sure the highest quality is produced each time. The critical specifications include height, diameter, neck thread and bottom thickness.
Septa are post-baked to provide the lowest levels of extractables in the industry. The softness (durometer) is measured for batch-to-batch reproducibility. All bonded septa are measured to certify bond strength reliability.
The caps are made from the finest grade of plastic available. The resin is tested for low levels of contamination. In addition, fit and dimension of the caps are measured to specification.