Tisk…
Graduated pipettes for enzyme test, Class AS
Dodavatel: NEUBERT VOLUME GLASSWAERE
0262-05-011 0262-07-011 0262-07-022 0262-07-051 0262-07-101
NEUB0262-05-011EA 0 CZK
NEUB0262-05-011 NEUB0262-07-011 NEUB0262-07-022 NEUB0262-07-051 NEUB0262-07-101
Graduated pipettes for enzyme test, Class AS
Pipety Pipety graduované
Graduated pipettes with long pulled out tip, ring divisions and partial sequence above O-point.
Certifikace: DIN 12699; Class AS, conformity certified.