Tisk…
Hand cleanser dispensers, Eurospender
Dodavatel: BODE CHEMIE
9814680 981467
BODE9814680EA 762 CZK
BODE9814680 BODE981467
Hand cleanser dispensers, Eurospender
Dávkovače mýdla a dávkovače mýdla
The Eurospender Safety plus is a wall dispenser for use with disposable disinfectant bottles.
  • Allows the use of containers from different manufacturers
  • Avoids microbial contamination of the pump head
  • Wide arm lever for easy and intuitive and hygienic elbow and arm operation
  • Integrated drip tray
  • Useable as table stand dispenser
  • Simple and fast bottle exchange
For hand disinfectants and wash lotions in funtional areas, personnal locks, examination rooms, wards, restrooms, medical practices, utility departments or hospital kitchens.