Tisk…

trans-β-Methylstyrene 99% (by GC), Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: trans-Propenylbenzene, trans-1-Phenyl-1-propene

M-Clarity™ quality level = MQ100

111848-10G 111848-1G
SIAL111848-10GEA 6650 CZK
SIAL111848-10G SIAL111848-1G
trans-β-Methylstyrene 99% (by GC), Sigma-Aldrich®
trans-beta-Methylstyrene

Trans-β-methylstyrene exhibits potential exothermic hazards at higher temperatures.


Used in the preparation of exo-chromans by the oxa analogue of the Povarov reaction and also in the epoxidation reaction of an oxidant solution with a solution of dichloromethane, where nickel is used as a catalyst and benzyltributylammonium bromide as a phase-transfer agent.

Vzorec: C₉H₁₀
MW: 118,18 g/mol
Bod varu: 175 °C (lit.)
Hustota: 0,911 g/ml (25 °C) (lit.)
Bod vzplanutí: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009280
CAS číslo: 873-66-5
UN: 2618
ADR: 3,III

Objednat

Výsledky specifikačního testu

Appearance (Colour) Colourless
Appearance (Form) Liquid
Infrared Spectrum Conforms to Structure
Purity (GC) ≥98.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR