Tisk…

β-D-Glucose ≥98%, crystalline solid

Dodavatel: Cayman Chemical

Synonyms: beta-D-Glucose

16775-10 16775-25
CAYM16775-10EA 1540 CZK
CAYM16775-10 CAYM16775-25
β-D-Glucose ≥98%, crystalline solid
β-D-Glucose
Vzorec: C₆H₁₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00063989
CAS číslo: 492-61-5

Objednat

Výsledky specifikačního testu

Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR